Brilliant Blue G, pure
Brilliant Blue G, pure
Brilliant Blue G, pure
Brilliant Blue G, pure
Thermo Scientific Chemicals
Brilliant Blue G, pure
CAS: 6104-58-1 | C47H48N3NaO7S2 | 854.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA191485000
Price (MYR)
2,823.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,823.00
EA
Add to cart
Specifications
Absorbance425 (at lambda min.)
Chemical Name or MaterialBrilliant blue G
Melting Point>100.0°C
CAS6104-58-1
Absorption Ratio0.95 to 1.15