Aluminum tri-sec-butoxide, 97%
Aluminum tri-sec-butoxide, 97%
Aluminum tri-sec-butoxide, 97%
Aluminum tri-sec-butoxide, 97%
Thermo Scientific Chemicals
Aluminum tri-sec-butoxide, 97%
CAS: 2269-22-9 | C12H27AlO3 | 246.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA191215000
Price (MYR)
730.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
730.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAluminum tri-sec-butoxide
CAS2269-22-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
If eye
View more
Lewis acid
RUO – Research Use Only