Osmium tetroxide, 99.9+%, (trace metal basis)
Osmium tetroxide, 99.9+%, (trace metal basis)
Osmium tetroxide, 99.9+%, (trace metal basis)
Osmium tetroxide, 99.9+%, (trace metal basis)
Thermo Scientific Chemicals
Osmium tetroxide, 99.9+%, (trace metal basis)
CAS: 20816-12-0 | O4Os | 254.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
250 mg
1 g
Catalog number FSA191185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Specifications
Boiling Point130.0°C
Chemical Name or MaterialOsmium tetroxide
Melting Point40.0°C to 42.0°C
CAS20816-12-0
ColorColorless to Yellow
View more
For ultra trace analysis, Oxidation reagent
RUO – Research Use Only