Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Ethyl cellulose, ethoxyl content 48%, 10 cps
Thermo Scientific Chemicals
Ethyl cellulose, ethoxyl content 48%, 10 cps
CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
Packaging:
Glass bottle
Catalog number FSA190965000
Price (MYR)
1,147.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,147.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl cellulose
ColorWhite to Yellow
Name NoteEthoxyl Content 48%, 10 Cps
CAS9004-57-3
Physical FormGranules or Crystalline Powder