Poly(methyl methacrylate), approx. M.W. 15000 (GPC)
Poly(methyl methacrylate), approx. M.W. 15000 (GPC)
Poly(methyl methacrylate), approx. M.W. 15000 (GPC)
Poly(methyl methacrylate), approx. M.W. 15000 (GPC)
Thermo Scientific Chemicals
Poly(methyl methacrylate), approx. M.W. 15000 (GPC)
CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
Packaging:
Plastic bottle
Catalog number FSA190691000
Price (MYR)
1,222.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,222.00
EA
Add to cart
Specifications
Chemical Name or MaterialPoly(methyl methacrylate)
ColorWhite
Name NoteAverage M.W. 15000 (GPC)
CAS9011-14-7
Physical FormBeads