Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA190522500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point4300.0°C
Chemical Name or MaterialZirconium(IV) oxide
Melting Point2700.0°C
Name Note98.5%
CAS1314-23-4