Litmus, pure, indicator grade
Litmus, pure, indicator grade
Thermo Scientific Chemicals
Litmus, pure, indicator grade
CAS: 1393-92-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA190221000
Price (MYR)
1,469.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,469.00
EA
Add to cart
Specifications
Chemical Name or MaterialLitmus
CAS1393-92-6
MDL NumberMFCD00131528
Merck Index15, 5603
Solubility InformationSolubility in water: soluble. Other solubilities: partially soluble in alcohol
View more
Indicator
RUO – Research Use Only