Victoria Blue B, pure
Victoria Blue B, pure
Victoria Blue B, pure
Victoria Blue B, pure
Thermo Scientific Chemicals
Victoria Blue B, pure
CAS: 2580-56-5 | C33H32ClN3 | 506.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA190190250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialVictoria blue B
Melting Point206.0°C
CAS2580-56-5
EINECS Number219-943-6
TSCATSCA