Tropaeolin O, pure
Tropaeolin O, pure
Tropaeolin O, pure
Thermo Scientific Chemicals
Tropaeolin O, pure
CAS: 547-57-9 | C12H10N2NaO5S | 317.271 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA190181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTropaeolin O
CAS547-57-9
RTECS NumberDB6145180
EINECS Number208-924-8
TSCATSCA
View more
Indicator
RUO – Research Use Only