Chrome Azurol S
Chrome Azurol S
Chrome Azurol S
Thermo Scientific Chemicals
Chrome Azurol S
CAS: 1667-99-8 | C23H13Cl2Na3O9S | 605.277 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA190051000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChrome Azurol S
CAS1667-99-8
Health Hazard 1Warning
Health Hazard 2GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
View more
Indicator
RUO – Research Use Only