Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Thermo Scientific Chemicals
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
CAS: 1310-53-8 | GeO2 | 104.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA190000500
Price (MYR)
3,061.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,061.00
EA
Add to cart
Specifications
Chemical Name or MaterialGermanium dioxide
Melting Point1115.0°C
CAS1310-53-8
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
For ultra trace analysis
RUO – Research Use Only