L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
Thermo Scientific Chemicals
L(+)-Lactic acid, 90% solution in water
CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA189872500
Price (MYR)
387.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
387.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL(+)-Lactic acid
CAS79-33-4
Boiling Point125.0°C
Health Hazard 1Danger
ColorColorless to Yellow