Cesium chloride, 99+%, for analysis
Cesium chloride, 99+%, for analysis
Cesium chloride, 99+%, for analysis
Cesium chloride, 99+%, for analysis
Thermo Scientific Chemicals
Cesium chloride, 99+%, for analysis
CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA189502500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point1290.0°C
Chemical Name or MaterialCesium chloride
Melting Point646.0°C
Name Notep.a.
CAS7647-17-8