Phloxine B, 85%, pure, high purity, Biological stain
Phloxine B, 85%, pure, high purity, Biological stain
Phloxine B, 85%, pure, high purity, Biological stain
Phloxine B, 85%, pure, high purity, Biological stain
Thermo Scientific Chemicals
Phloxine B, 85%, pure, high purity, Biological stain
CAS: 18472-87-2 | C20H2Br4Cl4Na2O5
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA189470250
Price (MYR)
832.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
832.00
BOT
Add to cart
Specifications
Absorbance875 to 1300 (at lambda max.)
Chemical Name or MaterialPhloxine B
CAS18472-87-2
Absorption Ratio1.38 to 2.24
Health Hazard 2GHS H Statement: