Sodium borohydride, 98+%, powder
Sodium borohydride, 98+%, powder
Sodium borohydride, 98+%, powder
Sodium borohydride, 98+%, powder
Thermo Scientific Chemicals
Sodium borohydride, 98+%, powder
CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
10 g
100 g
Packaging:
Plastic Bottle
Catalog number FSA189305000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium borohydride
Melting Point360.0°C
CAS16940-66-2
ColorWhite
Health Hazard 1Danger
View more
Reduction reagent
RUO – Research Use Only