4-(Trifluoromethyl)benzoic acid, 98%
4-(Trifluoromethyl)benzoic acid, 98%
4-(Trifluoromethyl)benzoic acid, 98%
4-(Trifluoromethyl)benzoic acid, 98%
Thermo Scientific Chemicals
4-(Trifluoromethyl)benzoic acid, 98%
CAS: 455-24-3 | C8H5F3O2 | 190.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA188460050
Price (MYR)
521.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
521.00
BOT
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)benzoic acid
Name Note98%
CAS455-24-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning