2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA188450050
Price (MYR)
393.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
393.00
BOT
Add to cart
Specifications
Chemical Name or Material2-(Trifluoromethyl)benzoic acid
Name Note98%
CAS433-97-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning