3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals
3-(Trifluoromethyl)benzaldehyde, 97%
CAS: 454-89-7 | C8H5F3O | 174.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA188440250
Price (MYR)
802.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
802.00
BOT
Add to cart
Specifications
Chemical Name or Material3-(Trifluoromethyl)benzaldehyde
Name Note97%
CAS454-89-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning