3-Fluoropyridine, 99%
3-Fluoropyridine, 99%
3-Fluoropyridine, 99%
3-Fluoropyridine, 99%
Thermo Scientific Chemicals
3-Fluoropyridine, 99%
CAS: 372-47-4 | C5H4FN | 97.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA188300050
Price (MYR)
1,402.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,402.00
EA
Add to cart
Specifications
Chemical Name or Material3-Fluoropyridine
CAS372-47-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0