2,2-Diphenylethanol, 97%
2,2-Diphenylethanol, 97%
2,2-Diphenylethanol, 97%
2,2-Diphenylethanol, 97%
Thermo Scientific Chemicals
2,2-Diphenylethanol, 97%
CAS: 1883-32-5 | C14H14O | 198.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
Packaging:
Glass bottle
Catalog number FSA188220020
Price (MYR)
333.00
EA
Add to cart
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
333.00
EA
Add to cart
Specifications
Chemical Name or Material2, 2-Diphenylethanol
CAS1883-32-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0