3-Furanmethanol, 99%
3-Furanmethanol, 99%
3-Furanmethanol, 99%
3-Furanmethanol, 99%
Thermo Scientific Chemicals
3-Furanmethanol, 99%
CAS: 4412-91-3 | C5H6O2 | 98.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA188080050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Furanmethanol
CAS4412-91-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0