Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Thermo Scientific Chemicals
Levamisole hydrochloride, 99+%
CAS: 16595-80-5 | C11H12N2S·ClH | 240.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA187870100
Price (MYR)
411.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
411.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLevamisole hydrochloride
CAS16595-80-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.