Ethylenediamine dihydrochloride, 98+%
Ethylenediamine dihydrochloride, 98+%
Ethylenediamine dihydrochloride, 98+%
Ethylenediamine dihydrochloride, 98+%
Thermo Scientific Chemicals
Ethylenediamine dihydrochloride, 98+%
CAS: 333-18-6 | C2H10Cl2N2 | 133.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA187655000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylenediamine dihydrochloride
Name Note98+%
CAS333-18-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: