Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
Thermo Scientific Chemicals
Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™
CAS: 13292-87-0 | C2H9BS | 75.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA186961000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13292-87-0, 108-88-3
Specifications
Chemical Name or MaterialBorane dimethyl sulfide complex
CAS13292-87-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
May cause drowsiness or dizziness.
May cause respiratory irritation.
Causes skin irritation
CAS108-88-3
View more
Reduction reagent
RUO – Research Use Only