Dimethyl isophthalate, 99%
Dimethyl isophthalate, 99%
Dimethyl isophthalate, 99%
Dimethyl isophthalate, 99%
Thermo Scientific Chemicals
Dimethyl isophthalate, 99%
CAS: 1459-93-4 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA186510025
Price (MYR)
1,113.00
BOT
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,113.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethyl isophthalate
Name Note99%
CAS1459-93-4
Melting Point>66.0°C