Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Oxalic acid, 98%, anhydrous
Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA186432500
Price (MYR)
331.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
331.00
EA
Add to cart
Specifications
Chemical Name or MaterialOxalic acid
CAS144-62-7
Melting Point189.0°C to 191.0°C
Health Hazard 1Danger
ColorWhite
View more
Suitable for battery materials development.
RUO – Research Use Only