Benzimidazole, 98%
Benzimidazole, 98%
Benzimidazole, 98%
Thermo Scientific Chemicals
Benzimidazole, 98%
CAS: 51-17-2 | C7H6N2 | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA186425000
Price (MYR)
1,249.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,249.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzimidazole
Name Note98%
CAS51-17-2
CAS Min %97.5
Health Hazard 1Warning