1,3-Dimethyl-2-imidazolidinone, 98%
1,3-Dimethyl-2-imidazolidinone, 98%
1,3-Dimethyl-2-imidazolidinone, 98%
1,3-Dimethyl-2-imidazolidinone, 98%
Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%
CAS: 80-73-9 | C5H10N2O | 114.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA185931000
Price (MYR)
330.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
330.00
BOT
Add to cart
Specifications
Chemical Name or Material1,3-Dimethyl-2-imidazolidinone
Name Note98%
CAS80-73-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger