Tetrabutylammonium bromide, 99+%
Tetrabutylammonium bromide, 99+%
Tetrabutylammonium bromide, 99+%
Tetrabutylammonium bromide, 99+%
Thermo Scientific Chemicals
Tetrabutylammonium bromide, 99+%
CAS: 1643-19-2 | C16H36BrN | 322.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
50 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA185685000
Price (MYR)
1,532.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,532.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetrabutylammonium bromide
Name Note99+%
CAS1643-19-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
View more
Phase transfer catalyst
RUO – Research Use Only