Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Thermo Scientific Chemicals
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
CAS: 12075-68-2 | C6H15Al2Cl3 | 247.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA185468000
Price (MYR)
4,544.00
ML
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
4,544.00
ML
Add to cart
Specifications
Chemical Name or MaterialEthylaluminium sesquichloride
CAS12075-68-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Suspected
CAS110-54-3
View more
Lewis acid
RUO – Research Use Only