Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Thermo Scientific Chemicals
Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number FSA185002500
Price (MYR)
723.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
723.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(acrylic acid)
Name Note50 wt. % Solution in Water
CAS9003-01-4
Physical FormLiquid
Health Hazard 1