Poly(acrylic acid), 63 wt% solution in water; approx. M.W. 2,000
Poly(acrylic acid), 63 wt% solution in water; approx. M.W. 2,000
Poly(acrylic acid), 63 wt% solution in water; approx. M.W. 2,000
Poly(acrylic acid), 63 wt% solution in water; approx. M.W. 2,000
Thermo Scientific Chemicals
Poly(acrylic acid), 63 wt% solution in water; approx. M.W. 2,000
CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
Packaging:
Glass bottle
Catalog number FSA184995000
Price (MYR)
1,599.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,599.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(acrylic acid)
Name Note63 wt. % Aqueous Solution
CAS9003-01-4
Physical FormLiquid
Boiling Point100.0°C