Deuterium oxide, for NMR, 100 atom % D, AcroSeal™
Deuterium oxide, for NMR, 100 atom % D, AcroSeal™
Deuterium oxide, for NMR, 100 atom % D, AcroSeal™
Deuterium oxide, for NMR, 100 atom % D, AcroSeal™
Deuterium oxide, for NMR, 100 atom % D, AcroSeal™
Thermo Scientific Chemicals
Deuterium oxide, for NMR, 100 atom % D, AcroSeal™
H2O20.03 g/mol
Have Questions?
Catalog NumberQuantity
FSA184761000100 mL
Catalog number FSA184761000
Price (MYR)
2,036.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
2,036.00
EA
Add to cart
Specifications
Quantity100 mL
Molecular FormulaH2O
MDL NumberMFCD00044636
InChI KeyXLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES[2H]O[2H]
View more
For NMR
RUO – Research Use Only