Glycerol, 99.5+%, for spectroscopy
Glycerol, 99.5+%, for spectroscopy
Glycerol, 99.5+%, for spectroscopy
Glycerol, 99.5+%, for spectroscopy
Thermo Scientific Chemicals
Glycerol, 99.5+%, for spectroscopy
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA184695000
Price (MYR)
578.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
578.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlycerol
Name Note99.5+%
CAS56-81-5
Absorbance(1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 340nm, 0.01 max. at 400nm, 0.02 max. at 320nm, 0.05 max. at 280nm, 0.25 max. at 225nm, 0.7 max. at 210nm, 1 max. at 206nm
Health Hazard 1HYGROSCOPIC
View more
For UV/VIS spectroscopy
RUO – Research Use Only