3,3'-Dimethoxybenzidine dihydrochloride, 98%
3,3'-Dimethoxybenzidine dihydrochloride, 98%
3,3'-Dimethoxybenzidine dihydrochloride, 98%
3,3'-Dimethoxybenzidine dihydrochloride, 98%
Thermo Scientific Chemicals
3,3'-Dimethoxybenzidine dihydrochloride, 98%
CAS: 20325-40-0 | C14H16N2O2·2HCl | 317.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA184470250
Price (MYR)
1,290.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,290.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 3'-Dimethoxybenzidine dihydrochloride
Name Note99%
CAS20325-40-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger