Isopropanol, 99.5%, for analysis
Isopropanol, 99.5%, for analysis
Isopropanol, 99.5%, for analysis
Isopropanol, 99.5%, for analysis
Thermo Scientific Chemicals
Isopropanol, 99.5%, for analysis
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA184130025
Price (MYR)
848.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
848.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
Name NoteReag. Ph. Eur.
Absorbance0.01 max. (EurPh) at 310nm, 0.02 max. (EurPh) at 290nm, 0.03 max. (EurPh) at 270nm, 0.1 max. (EurPh) at 250nm, 0.3 max. (EurPh) at 230nm
CAS67-63-0
View more
For analysis
RUO – Research Use Only