tert-Butylchlorodimethylsilane, 98%
tert-Butylchlorodimethylsilane, 98%
tert-Butylchlorodimethylsilane, 98%
tert-Butylchlorodimethylsilane, 98%
Thermo Scientific Chemicals
tert-Butylchlorodimethylsilane, 98%
CAS: 18162-48-6 | C6H15ClSi | 150.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Glass bottle
Catalog number FSA183930250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtert-Butylchlorodimethylsilane
CAS18162-48-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha