1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
1H-Benzotriazole, 99%
Thermo Scientific Chemicals
1H-Benzotriazole, 99%
CAS: 95-14-7 | C6H5N3 | 119.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA183885000
Price (MYR)
844.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
844.00
BOT
Add to cart
Specifications
Chemical Name or Material1H-Benzotriazole
Name Note98%
CAS95-14-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
View more
Indicator
RUO – Research Use Only