3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
Thermo Scientific Chemicals
3-Phenoxybenzyl alcohol, 98%
CAS: 13826-35-2 | C13H12O2 | 200.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA183770250
Price (MYR)
678.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
678.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Phenoxybenzyl alcohol
Name Note98%
CAS13826-35-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning