Poly(vinyl acetate), approx. M.W. 170,000
Poly(vinyl acetate), approx. M.W. 170,000
Poly(vinyl acetate), approx. M.W. 170,000
Poly(vinyl acetate), approx. M.W. 170,000
Thermo Scientific Chemicals
Poly(vinyl acetate), approx. M.W. 170,000
CAS: 9003-20-7 | (C4H6O2)n | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA183255000
Price (MYR)
1,080.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,080.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPoly(vinyl acetate)
Name NoteAverage M.W. 170.000
CAS9003-20-7
Physical FormBeads
RTECS NumberAK0920000