Poly(caprolactone) diol, average M.N. 2000
Poly(caprolactone) diol, average M.N. 2000
Poly(caprolactone) diol, average M.N. 2000
Poly(caprolactone) diol, average M.N. 2000
Thermo Scientific Chemicals
Poly(caprolactone) diol, average M.N. 2000
CAS: 36890-68-3 | (C6H10O2)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA183215000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(caprolactone) diol
ColorWhite
CAS36890-68-3
Physical FormWaxy Solid
Molecular Formula(C6H10O2)n