Poly(vinyl alcohol), 99.3-100.0% hydrolyzed,M.W. appr. 146,000-186,000
Poly(vinyl alcohol), 99.3-100.0% hydrolyzed,M.W. appr. 146,000-186,000
Poly(vinyl alcohol), 99.3-100.0% hydrolyzed,M.W. appr. 146,000-186,000
Poly(vinyl alcohol), 99.3-100.0% hydrolyzed,M.W. appr. 146,000-186,000
Thermo Scientific Chemicals
Poly(vinyl alcohol), 99.3-100.0% hydrolyzed,M.W. appr. 146,000-186,000
Indicator | CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Catalog number FSA183121000
Price (MYR)
234.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
234.00
EA
Add to cart
Specifications
Chemical Name or MaterialPoly(vinyl alcohol)
CAS9002-89-5
Melting Point230.0°C to 240.0°C
pH5.0 to 7.0 (4% aq. soln.)
Infrared SpectrumAuthentic