Check Order Status
Poxygrid Lead Flask Ring Holder
Actual product may vary
Thermo Scientific Chemicals
Poxygrid Lead Flask Ring Holder
Wire formed, epoxy coated holder keeps lead ring weights organized and handy
Have Questions?
Catalog number
FSA183071000
Price (MYR)
397.00
BOT
Add to cart
Price (MYR)
397.00
BOT
Add to cart
Small footprint; circular base is only 19cm (7½ in.) in diameter
Rings having an inside diameter of 17.8mm (0.7 in.) or larger stack neatly on a tapered center post (17.1cm (6¾ in.) high)
Steam autoclavable at 121°C (250°F)
Specifications
Color
Blue
Diameter (Metric) Base
19 cm
Material
Epoxy Coated Wire
Height (Metric)
17.1 cm
No. per Pack
1
Autoclavable
Autoclavable