p-Toluenesulfonyl isocyanate, 96%
p-Toluenesulfonyl isocyanate, 96%
p-Toluenesulfonyl isocyanate, 96%
p-Toluenesulfonyl isocyanate, 96%
Thermo Scientific Chemicals
p-Toluenesulfonyl isocyanate, 96%
CAS: 4083-64-1 | C8H7NO3S | 197.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA183070250
Price (MYR)
396.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
396.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Toluenesulfonyl isocyanate
CAS4083-64-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Reacts violently with water.
Health Hazard 3GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.