4-Hydroxyphenethyl alcohol, 98%
4-Hydroxyphenethyl alcohol, 98%
4-Hydroxyphenethyl alcohol, 98%
4-Hydroxyphenethyl alcohol, 98%
4-Hydroxyphenethyl alcohol, 98%
CAS: 501-94-0 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA182500050
Price (MYR)
525.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
525.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Hydroxyphenethyl alcohol
Name Note98%
CAS501-94-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning