o-Cresolphthalein Complexone, pure
o-Cresolphthalein Complexone, pure
o-Cresolphthalein Complexone, pure
o-Cresolphthalein Complexone, pure
Thermo Scientific Chemicals
o-Cresolphthalein Complexone, pure
CAS: 2411-89-4 | C32H32N2O12 | 636.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA182440250
Price (MYR)
1,254.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,254.00
BOT
Add to cart
Specifications
Chemical Name or Materialo-Cresolphthalein Complexone
Melting Point181.0°C
Name NotePure
CAS2411-89-4
EINECS Number219-318-8
View more
Indicator
RUO – Research Use Only