Methyl cellulose, viscosity 15 cP (2% solution in water)
Methyl cellulose, viscosity 15 cP (2% solution in water)
Methyl cellulose, viscosity 15 cP (2% solution in water)
Methyl cellulose, viscosity 15 cP (2% solution in water)
Thermo Scientific Chemicals
Methyl cellulose, viscosity 15 cP (2% solution in water)
CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA182312500
Price (MYR)
618.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
618.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl cellulose
ColorGray-White
Name NoteViscosity (2% Solution in Water) 12-18 CP
CAS9004-67-5
Physical FormPowder