2-Methoxyphenethylamine, 98%
2-Methoxyphenethylamine, 98%
2-Methoxyphenethylamine, 98%
2-Methoxyphenethylamine, 98%
Thermo Scientific Chemicals
2-Methoxyphenethylamine, 98%
CAS: 2045-79-6 | C9H13NO | 151.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA182170050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methoxyphenethylamine
CAS2045-79-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0