Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Thermo Scientific Chemicals
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, >-99.85%, C4H8O, CAS Number-109-99-9, 128-37-0, tetraidrofurano, furanidine, tetrahydrofuranne, butylene oxide, tetrahydrofuran, furan, tetrahydro, oxacyclopentane, hydrofuran, 1,4-epoxybutane, tetramethylene oxide, 1L, 99.9%, 17, 10, CHEBI:26911, Undesignated, 0.8800g/mL | CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass Bottle
Catalog number FSA181500025
Price (MYR)
1,314.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,314.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetrahydrofuran
Name NoteAnhydrous, Stabilized
CAS109-99-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.