N,N-Dimethyl-p-phenylenediamine sulfate, 99%
N,N-Dimethyl-p-phenylenediamine sulfate, 99%
N,N-Dimethyl-p-phenylenediamine sulfate, 99%
N,N-Dimethyl-p-phenylenediamine sulfate, 99%
Thermo Scientific Chemicals
N,N-Dimethyl-p-phenylenediamine sulfate, 99%
CAS: 536-47-0 | C8H14N2O4S | 234.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA181431000
Price (MYR)
2,265.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,265.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethyl-p-phenylenediamine sulfate
Name Note99%
CAS536-47-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger